Learn More
Agilent Technologies Microplate Lid, Universal, Black, Polystyrene, 100/Pk.
Catalog No. 50995884
Encompass
These universal black polystyrene microplate lids offer a light-sensitive sealing solution for laboratory microplate applications. Engineered for broad compatibility with most microplate formats, these lids feature a robot-friendly design for reliable integration into automated workflows.
  • Material: Polystyrene
  • Color: Black
  • Compatibility: Universal fit for most microplates
  • Design: Robot-friendly
  • Quantity: 100/pk
Catalog No. 50-995-884 Supplier Agilent Technologies Supplier No. 200858100
These universal black polystyrene microplate lids offer a light-sensitive sealing solution for laboratory microplate applications. Engineered for broad compatibility with most microplate formats, these lids feature a robot-friendly design for reliable integration into automated workflows.
  • Material: Polystyrene
  • Color: Black
  • Compatibility: Universal fit for most microplates
  • Design: Robot-friendly
  • Quantity: 100/pk