Learn More
Agilent Technologies Universal Microplate Lids, Clear, Polystyrene, 100/Pk.
Catalog No. 50995883
Encompass
These universal microplate lids are designed for use with most microplates to prevent contamination and sample evaporation. Constructed from clear polystyrene resin, these covers provide excellent clarity for visual inspection of contents while maintaining a secure fit.
  • Manufactured from clear polystyrene resin
  • Universal size fits most microplates
  • Robot-friendly design
  • Specifically designed for optimal fit to square-well microplates
  • Features low permeability to alcohols
Catalog No. 50-995-883 Supplier Agilent Technologies Supplier No. 200856100
These universal microplate lids are designed for use with most microplates to prevent contamination and sample evaporation. Constructed from clear polystyrene resin, these covers provide excellent clarity for visual inspection of contents while maintaining a secure fit.
  • Manufactured from clear polystyrene resin
  • Universal size fits most microplates
  • Robot-friendly design
  • Specifically designed for optimal fit to square-well microplates
  • Features low permeability to alcohols