Learn More
Euthanex E-098 Lid Stem for Euthanex Lids.
Catalog No. NC1553328
Encompass
E-098 Lid Stem comes with every Euthanex Lid (lid sold separately). The Lid Stem is sold as a replacement part for your existing lid system.The Lid Stem threads into the middle hole of the lid and allows gas to flow when connected to a Euthanex regulator, flow meter, or EP-1130 Connection Hose.
Catalog No. NC1553328 Supplier Euthanex Supplier No. E098
E-098 Lid Stem comes with every Euthanex Lid (lid sold separately). The Lid Stem is sold as a replacement part for your existing lid system.The Lid Stem threads into the middle hole of the lid and allows gas to flow when connected to a Euthanex regulator, flow meter, or EP-1130 Connection Hose.