Learn More
Boekel Scientific™ Lid for Thermal Mixers
GREENER_CHOICESDP
Catalog No. 15600350 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
270800 and 270900 Thermal Mixers
Catalog No. For Use With (Equipment)
15-600-350 270800 and 270900 Thermal Mixers
1 options
Catalog No. 15-600-350
Only null left
Add to Cart
Add to Cart
This lid will help ensure the optimum stability and uniformity throughout heating and cooling and increase reliability and reproducibility.
  • Included with 270818 thermal block
Specifications
Product Type Thermal Mixer Block Lid
For Use With (Equipment) 270800 and 270900 Thermal Mixers