Learn More
Medchemexpress LLC HY-101815 1mg Medchemexpress, Lidanserin CAS:73725-85-6 Purity:>98%
SDP
Catalog No. 501932221
Encompass_Preferred
Medchemexpress, HY-101815 1mg Lidanserin CAS:73725-85-6 Lidanserin is a drug which acts as a combined 5-HT 2A and α 1 -adrenergic receptor antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2221 Supplier Medchemexpress LLC Supplier No. HY1018151MG
Medchemexpress, HY-101815 1mg Lidanserin CAS:73725-85-6 Lidanserin is a drug which acts as a combined 5-HT 2A and α 1 -adrenergic receptor antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.