Learn More
Med Vet International RX Lidocaine with Epinephrine, 50mL, Multi-Dose, 25/Pack
Catalog No. 503045324
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Lidocaine Hydrochloride and Epinephrine Injection, USP is indicated for production of local or regional anesthesia by infiltration techniques such as percutaneous injection. Lidocaine stabilizes the neuronal membrane by inhibiting the ionic fluxes required for the initiation and conduction of nerve impulses, thereby effecting local anesthetic action. Lidocaine HCL 2% Epinephrine: 1:100,000
Catalog No. 50-304-5324 Supplier Med Vet International Supplier No. RX00409318203
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Lidocaine Hydrochloride and Epinephrine Injection, USP is indicated for production of local or regional anesthesia by infiltration techniques such as percutaneous injection. Lidocaine stabilizes the neuronal membrane by inhibiting the ionic fluxes required for the initiation and conduction of nerve impulses, thereby effecting local anesthetic action. Lidocaine HCL 2% Epinephrine: 1:100,000