Learn More
Med Vet International Lidocaine Hcl 2%, 100 mL Injection, Vet Use
Catalog No. 502828673
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Description: Lidocaine is a potent local anesthetic for producing epidural, nerve conduction and infiltration anesthesia. For use in dogs, cats, cattle and horses. Each mL of sterile aqueous solution contains: -Lidocaine Hydrochloride 2.0% -Propylene Glycol 5.2% -Sodium Chloride 0.5% -Sodium Lactate 0.5% -Methylparaben 0.15%, Sodium Metabisulfite 0.10%, -Propylparaben 0.03% and Disodium Edetate 0.001% as preservatives.
Catalog No. 50-282-8673 Supplier Med Vet International Supplier No. RXLIDO100INJ
 Restricted Item
Add to Cart
Add to Cart
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Description: Lidocaine is a potent local anesthetic for producing epidural, nerve conduction and infiltration anesthesia. For use in dogs, cats, cattle and horses. Each mL of sterile aqueous solution contains: -Lidocaine Hydrochloride 2.0% -Propylene Glycol 5.2% -Sodium Chloride 0.5% -Sodium Lactate 0.5% -Methylparaben 0.15%, Sodium Metabisulfite 0.10%, -Propylparaben 0.03% and Disodium Edetate 0.001% as preservatives.