Learn More
Cayman Chemical Lidocaine-d10, Purity: ≥99% deuterated forms (d1-d10), CAS: 851528-09-1, 10 mg
SDP
Catalog No. 502911476
Encompass_Preferred
An internal standard for the quantification of lidocaine by GC- or LC-MS
Catalog No. 50-291-1476 Supplier Cayman Chemical Supplier No. 3506610 MG
An internal standard for the quantification of lidocaine by GC- or LC-MS