Learn More
Apexbio Technology LLC Lidocaine, 10g Cas# 137-58-6
SDP
Supplier:  Apexbio Technology LLC A145010000
Encompass_Preferred
Anasthetic and class Ib antiarrhythmic agent.?Blocks voltage-gated sodium channels in the inactivated state. Other sizes are available. Please inqury us for quote.
Catalog No. 50-199-8511