Learn More
Medchemexpress LLC Lidocaine | 137-58-6 | 99.96% | 234.34 | 5 G
SDP
Catalog No. 5000366020
Encompass_Preferred
Lidocaine inhibits sodium channels involving complex voltage and dependence. It decreases growth, migration, and invasion of gastric carcinoma cells by up-regulating miR-145 expression and further inactivating MEK/ERK and NF-κB signaling pathways. As an amide derivative, it has potential for ventricular arrhythmia research.
  • Inhibits sodium channels
  • Decreases growth, migration, and invasion of gastric carcinoma cells
  • Up-regulates miR-145 expression
  • Inactivates MEK/ERK and NF-κB signaling pathways
  • Causes reversible tail nerve block in rats
  • Inhibits human TREK-1 in HEK293 cells
  • Inhibits cell viability in human gastric cancer cell lines
  • Increases apoptotic cell rate
  • Down-regulates Cyclin D1 and up-regulates p21 expression
Catalog No. 50-003-66020 Supplier Medchemexpress LLC Supplier No. 5GHYB0185
Lidocaine inhibits sodium channels involving complex voltage and dependence. It decreases growth, migration, and invasion of gastric carcinoma cells by up-regulating miR-145 expression and further inactivating MEK/ERK and NF-κB signaling pathways. As an amide derivative, it has potential for ventricular arrhythmia research.
  • Inhibits sodium channels
  • Decreases growth, migration, and invasion of gastric carcinoma cells
  • Up-regulates miR-145 expression
  • Inactivates MEK/ERK and NF-κB signaling pathways
  • Causes reversible tail nerve block in rats
  • Inhibits human TREK-1 in HEK293 cells
  • Inhibits cell viability in human gastric cancer cell lines
  • Increases apoptotic cell rate
  • Down-regulates Cyclin D1 and up-regulates p21 expression