Learn More
Apexbio Technology LLC Lidocaine 137-58-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062777
Encompass_Preferred
Lidocaine (CAS 137-58-6) is a small-molecule anesthetic and class Ib antiarrhythmic agent commonly employed in biomedical research to investigate neuronal excitability and cardiac electrophysiology It exerts its biological activity primarily by binding preferentially to voltage-gated sodium channels in their inactivated conformational state thereby inhibiting sodium ion influx This blockage suppresses action potential propagation and decreases neuronal and cardiac tissue excitability Lidocaine is frequently utilized experimentally to study sodium channel function and pathophysiology of arrhythmias
Catalog No. 50-000-62777 Supplier Apexbio Technology LLC Supplier No. A14505.1
Lidocaine (CAS 137-58-6) is a small-molecule anesthetic and class Ib antiarrhythmic agent commonly employed in biomedical research to investigate neuronal excitability and cardiac electrophysiology It exerts its biological activity primarily by binding preferentially to voltage-gated sodium channels in their inactivated conformational state thereby inhibiting sodium ion influx This blockage suppresses action potential propagation and decreases neuronal and cardiac tissue excitability Lidocaine is frequently utilized experimentally to study sodium channel function and pathophysiology of arrhythmias