Learn More
Medchemexpress LLC HY-B0185 500mg Medchemexpress, Lidocaine CAS:137-58-6 Purity:>98%
SDP
Catalog No. 502082402
Encompass_Preferred
Medchemexpress, HY-B0185 500mg Lidocaine CAS:137-58-6 Lidocaine (Lignocaine) inhibits sodium channels involving complex voltage and using dependence. Lidocaine decreases growth, migration and invasion of gastric carcinoma cells via up-regulating miR-145 expression and further inactivation of MEK/ERK and NF-κB signaling pathways. Lidocaine is an amide derivative commonly used to anesthetize. Lidocaine is a drug to treat ventricular arrhythmia and an effective tumor-inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2402 Supplier Medchemexpress LLC Supplier No. HYB0185500MG
Medchemexpress, HY-B0185 500mg Lidocaine CAS:137-58-6 Lidocaine (Lignocaine) inhibits sodium channels involving complex voltage and using dependence. Lidocaine decreases growth, migration and invasion of gastric carcinoma cells via up-regulating miR-145 expression and further inactivation of MEK/ERK and NF-κB signaling pathways. Lidocaine is an amide derivative commonly used to anesthetize. Lidocaine is a drug to treat ventricular arrhythmia and an effective tumor-inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.