Learn More
Medchemexpress LLC Lidocaine-d10 | 851528-09-1 | 98.9% | 244.40 g/mol | C14H12D10N2O | 10 MG
SDP
Catalog No. 5000298839
Encompass_Preferred
Deuterium-labeled lidocaine (lidocaine-d10) supplied as an off-white to light yellow solid for use as a tracer or internal standard in quantitative analytical methods. It provides a mass-shifted analogue of lidocaine suitable for NMR, GC-MS, and LC-MS workflows, and is provided in small pack sizes for laboratory use.
  • Deuterium-labeled (d10) internal standard for mass spectrometry and NMR
  • High purity (~98.9%) suitable for analytical quantitation
  • Solid powder, off-white to light yellow, soluble in DMSO at 100 mg/mL
  • Storage: powder at -20°C or 4°C depending on shelf-life requirements
  • Suitable as a tracer and for pharmacokinetic or method development studies
Catalog No. 50-002-98839 Supplier Medchemexpress LLC Supplier No. HYB0185S110MG
Deuterium-labeled lidocaine (lidocaine-d10) supplied as an off-white to light yellow solid for use as a tracer or internal standard in quantitative analytical methods. It provides a mass-shifted analogue of lidocaine suitable for NMR, GC-MS, and LC-MS workflows, and is provided in small pack sizes for laboratory use.
  • Deuterium-labeled (d10) internal standard for mass spectrometry and NMR
  • High purity (~98.9%) suitable for analytical quantitation
  • Solid powder, off-white to light yellow, soluble in DMSO at 100 mg/mL
  • Storage: powder at -20°C or 4°C depending on shelf-life requirements
  • Suitable as a tracer and for pharmacokinetic or method development studies