Learn More
Medchemexpress LLC Lidocaine-d10 hydrochloride | 1189959-13-4 | 99.6% | 5 MG
SDP
Catalog No. 5000407003
Encompass_Preferred
Lidocaine-d10 hydrochloride is the deuterium-labeled Lidocaine hydrochloride. It inhibits sodium channels involving complex voltage and use dependence, and decreases the growth, migration, and invasion of gastric carcinoma cells by up-regulating miR-145 expression and inactivating MEK/ERK and NF-κB signaling pathways. This amide derivative has potential for research into ventricular arrhythmia and is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-07003 Supplier Medchemexpress LLC Supplier No. HYB0185AS5MG
Add to Cart
Add to Cart
Lidocaine-d10 hydrochloride is the deuterium-labeled Lidocaine hydrochloride. It inhibits sodium channels involving complex voltage and use dependence, and decreases the growth, migration, and invasion of gastric carcinoma cells by up-regulating miR-145 expression and inactivating MEK/ERK and NF-κB signaling pathways. This amide derivative has potential for research into ventricular arrhythmia and is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.