Learn More
Medchemexpress LLC Lidocaine-d10 hydrochloride | 1189959-13-4 | MFCD08705335 | 99.6% | 280.86 g/mol | C14H13D10ClN2O | 10 MG
SDP
Catalog No. 5000298945
Encompass_Preferred
Lidocaine-d10 hydrochloride is the deuterium-labeled hydrochloride salt of lidocaine supplied for research use. It is intended as an internal standard or tracer in analytical chemistry, mass spectrometry, and pharmacokinetic studies, and for research into sodium channel modulation.
  • Deuterium-labeled hydrochloride salt for use as an internal standard.
  • Molecular weight 280.86 g/mol; CAS number 1189959-13-4.
  • High purity (≈99.6%) suitable for analytical applications.
  • Available in small pack sizes for reference standards, including 10 mg.
  • Store powder at -20°C for long-term stability; in solvent store at -80°C for extended storage.
Catalog No. 50-002-98945 Supplier Medchemexpress LLC Supplier No. HYB0185AS10MG
Lidocaine-d10 hydrochloride is the deuterium-labeled hydrochloride salt of lidocaine supplied for research use. It is intended as an internal standard or tracer in analytical chemistry, mass spectrometry, and pharmacokinetic studies, and for research into sodium channel modulation.
  • Deuterium-labeled hydrochloride salt for use as an internal standard.
  • Molecular weight 280.86 g/mol; CAS number 1189959-13-4.
  • High purity (≈99.6%) suitable for analytical applications.
  • Available in small pack sizes for reference standards, including 10 mg.
  • Store powder at -20°C for long-term stability; in solvent store at -80°C for extended storage.