Learn More
Medchemexpress LLC Lidocaine-d6 hydrochloride | 2517378-96-8 | >95% | 1 MG
SDP
Catalog No. 5000407425
Encompass_Preferred
Lidocaine-d6 hydrochloride is a deuterium-labeled form of Lidocaine hydrochloride, primarily used in research. This compound acts as a sodium channel inhibitor and has shown potential in reducing the growth, migration, and invasion of gastric carcinoma cells by up-regulating miR-145 expression and inactivating MEK/ERK and NF-κB signaling pathways. It is also recognized as an effective agent for treating ventricular arrhythmia.
  • Used as a tracer for research applications
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Inhibits sodium channels
  • Decreases growth, migration, and invasion of gastric carcinoma cells
  • Acts as an agent to treat ventricular arrhythmia
Catalog No. 50-004-07425 Supplier Medchemexpress LLC Supplier No. HYB0185AS11MG
Add to Cart
Add to Cart
Lidocaine-d6 hydrochloride is a deuterium-labeled form of Lidocaine hydrochloride, primarily used in research. This compound acts as a sodium channel inhibitor and has shown potential in reducing the growth, migration, and invasion of gastric carcinoma cells by up-regulating miR-145 expression and inactivating MEK/ERK and NF-κB signaling pathways. It is also recognized as an effective agent for treating ventricular arrhythmia.
  • Used as a tracer for research applications
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Inhibits sodium channels
  • Decreases growth, migration, and invasion of gastric carcinoma cells
  • Acts as an agent to treat ventricular arrhythmia