Learn More
Enzo Life Sciences Lidocaine . hydrochloride. hydrate (5g). CAS: 6108-05-0
SDP
Catalog No. 502009509
Encompass_Preferred
Na+ channel blocker. Alternative name: 2-Diethylamino-N-(2',6'-dimethylphenyl)acetamide . HCl. Purity: ≥98% (Assay). Solubility: Soluble in water. Long Term Storage: Ambient.
Catalog No. 50-200-9509 Supplier Enzo Life Sciences Supplier No. ALX550251G005
Na+ channel blocker. Alternative name: 2-Diethylamino-N-(2',6'-dimethylphenyl)acetamide . HCl. Purity: ≥98% (Assay). Solubility: Soluble in water. Long Term Storage: Ambient.