Learn More
Medchemexpress LLC Lidocaine hydrochloride | 73-78-9 | MFCD00034865 | 99.84% | 10 G
SDP
Catalog No. 501639243
Encompass_Preferred
Lidocaine hydrochloride is a sodium channel inhibitor that exhibits complex voltage and use dependence. It reduces the growth, migration, and invasion of gastric carcinoma cells by up-regulating miR-145 expression and subsequently inactivating the MEK/ERK and NF-κB signaling pathways. This amide derivative serves as a treatment for ventricular arrhythmia and is an effective tumor inhibitor.
  • Inhibits sodium channels with voltage and use dependence.
  • Decreases growth, migration, and invasion of gastric carcinoma cells.
  • Up-regulates miR-145 expression and inactivates MEK/ERK and NF-κB signaling pathways.
  • Serves as a treatment for ventricular arrhythmia.
  • Effective tumor inhibitor.
Catalog No. 50-163-9243 Supplier Medchemexpress LLC Supplier No. HYB0185A10G
Lidocaine hydrochloride is a sodium channel inhibitor that exhibits complex voltage and use dependence. It reduces the growth, migration, and invasion of gastric carcinoma cells by up-regulating miR-145 expression and subsequently inactivating the MEK/ERK and NF-κB signaling pathways. This amide derivative serves as a treatment for ventricular arrhythmia and is an effective tumor inhibitor.
  • Inhibits sodium channels with voltage and use dependence.
  • Decreases growth, migration, and invasion of gastric carcinoma cells.
  • Up-regulates miR-145 expression and inactivates MEK/ERK and NF-κB signaling pathways.
  • Serves as a treatment for ventricular arrhythmia.
  • Effective tumor inhibitor.