Learn More
Medchemexpress LLC Lidocaine hydrochloride | 73-78-9 | 99.8% | 5 G
SDP
Catalog No. 5000406636
Encompass_Preferred
Lidocaine hydrochloride (Lignocaine hydrochloride) is an amide derivative that inhibits sodium channels. It decreases the growth, migration, and invasion of gastric carcinoma cells by up-regulating miR-145 expression and inactivating the MEK/ERK and NF-κB signaling pathways. It is used to treat ventricular arrhythmia and is an effective tumor-inhibitor.
  • Decreases cell proliferation
  • Inhibits cell viability
  • Increases the apoptotic cell rate
  • Down-regulates Cyclin D1 and up-regulates p21 expression
  • Causes completely reversible tail nerve block in rats
Catalog No. 50-004-06636 Supplier Medchemexpress LLC Supplier No. HYB0185A5G
Lidocaine hydrochloride (Lignocaine hydrochloride) is an amide derivative that inhibits sodium channels. It decreases the growth, migration, and invasion of gastric carcinoma cells by up-regulating miR-145 expression and inactivating the MEK/ERK and NF-κB signaling pathways. It is used to treat ventricular arrhythmia and is an effective tumor-inhibitor.
  • Decreases cell proliferation
  • Inhibits cell viability
  • Increases the apoptotic cell rate
  • Down-regulates Cyclin D1 and up-regulates p21 expression
  • Causes completely reversible tail nerve block in rats