Learn More
Medchemexpress LLC Lidocaine hydrochloride hydrate | 6108-05-0 | 99.8% | 100 MG
SDP
Catalog No. 5000407428
Encompass_Preferred
Lidocaine hydrochloride hydrate inhibits sodium channels involving complex voltage and using dependence. It decreases growth, migration, and invasion of gastric carcinoma cells via up-regulating miR-145 expression and further inactivation of MEK/ERK and NF-κB signaling pathways. It is an amide derivative with potential for the research of ventricular arrhythmia.
  • Inhibits sodium channels
  • Decreases growth, migration, and invasion of gastric carcinoma cells
  • Up-regulates miR-145 expression
  • Inactivates MEK/ERK and NF-κB signaling pathways
  • Potential for research of ventricular arrhythmia
  • Causes completely reversible tail nerve block in rats
  • Decreases cell proliferation in human gastric cancer cell line MKN45
  • Inhibits MKN45 cell viability
  • Increases apoptotic cell rate in MKN45 cells
  • Down-regulates Cyclin D1 and up-regulates p21 expression
Catalog No. 50-004-07428 Supplier Medchemexpress LLC Supplier No. HYB0185B100MG
Lidocaine hydrochloride hydrate inhibits sodium channels involving complex voltage and using dependence. It decreases growth, migration, and invasion of gastric carcinoma cells via up-regulating miR-145 expression and further inactivation of MEK/ERK and NF-κB signaling pathways. It is an amide derivative with potential for the research of ventricular arrhythmia.
  • Inhibits sodium channels
  • Decreases growth, migration, and invasion of gastric carcinoma cells
  • Up-regulates miR-145 expression
  • Inactivates MEK/ERK and NF-κB signaling pathways
  • Potential for research of ventricular arrhythmia
  • Causes completely reversible tail nerve block in rats
  • Decreases cell proliferation in human gastric cancer cell line MKN45
  • Inhibits MKN45 cell viability
  • Increases apoptotic cell rate in MKN45 cells
  • Down-regulates Cyclin D1 and up-regulates p21 expression