Learn More
Medchemexpress LLC Lidocaine hydrochloride hydrate | 6108-05-0 | 99.8% | 50 MG
SDP
Catalog No. 5000405767
Encompass_Preferred
Lidocaine hydrochloride hydrate is a sodium channel blocker that inhibits sodium channels with complex voltage and use dependence. It decreases the growth, migration, and invasion of gastric carcinoma cells by up-regulating miR-145 expression and inactivating the MEK/ERK and NF-κB signaling pathways. This amide derivative shows potential for research into ventricular arrhythmia.
  • Inhibits sodium channels
  • Decreases growth, migration, and invasion of gastric carcinoma cells
  • Upregulates miR-145 expression
  • Inactivates MEK/ERK and NF-κB signaling pathways
  • Decreases cell proliferation and viability in MKN45 cells
  • Increases apoptotic cell rate in MKN45 cells
  • Down-regulates Cyclin D1 and up-regulates p21 expression in MKN45 cells
Catalog No. 50-004-05767 Supplier Medchemexpress LLC Supplier No. HYB0185B50MG
Lidocaine hydrochloride hydrate is a sodium channel blocker that inhibits sodium channels with complex voltage and use dependence. It decreases the growth, migration, and invasion of gastric carcinoma cells by up-regulating miR-145 expression and inactivating the MEK/ERK and NF-κB signaling pathways. This amide derivative shows potential for research into ventricular arrhythmia.
  • Inhibits sodium channels
  • Decreases growth, migration, and invasion of gastric carcinoma cells
  • Upregulates miR-145 expression
  • Inactivates MEK/ERK and NF-κB signaling pathways
  • Decreases cell proliferation and viability in MKN45 cells
  • Increases apoptotic cell rate in MKN45 cells
  • Down-regulates Cyclin D1 and up-regulates p21 expression in MKN45 cells