Learn More
Specifications
Specifications
| Quantity | 10 g |
| IUPAC Name | 2-(diethylamino)-N-(2,6-dimethylphenyl)acetamide;hydrate;hydrochloride |
| Formula Weight | 270.8 |
| Grade | Not Available |
| Physical Form | Powder |
| Chemical Name or Material | Lidocaine Hydrochloride |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.