Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lidocaine hydrochloride, MP Biomedicals™

Catalog No. ICN19391780 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19391780 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19391780 Supplier MP Biomedicals, Inc Supplier No. 0219391780
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6108-05-0
Molecular Formula C14H25ClN2O2
Molecular Weight (g/mol) 288.82
MDL Number MFCD00150329
InChI Key YECIFGHRMFEPJK-UHFFFAOYSA-N
Synonym lidocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride hydrate, dolicaine, linocaine hydrochloride, lidocaine hydrochloride hydrate, xylocaine hydrochloride monohydrate, lignocaine hydrochloride monohydrate, 2-diethylamino-n-2,6-dimethylphenyl acetamide hydrochloride monohydrate, zingo tm, lidocaine hcl hydrate
PubChem CID 16219577
ChEBI CHEBI:60791
IUPAC Name 2-(diethylamino)-N-(2,6-dimethylphenyl)acetamide;hydrate;hydrochloride
SMILES O.[Cl-].CC[NH+](CC)CC(=O)NC1=C(C)C=CC=C1C

Specifications

Color White
Quantity 100 g
Formula Weight 270.8
Physical Form Crystalline Powder
Chemical Name or Material Lidocaine hydrochloride
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.