Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lidocaine, MP Biomedicals

Catalog No. ICN19011125 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19011125 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19011125 Supplier MP Biomedicals, Inc Supplier No. 0219011125
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium channel blocker and class IB antiarrhythmic agent.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-58-6
Molecular Formula C14H22N2O
Molecular Weight (g/mol) 234.343
InChI Key NNJVILVZKWQKPM-UHFFFAOYSA-N
Synonym lidocaine, lignocaine, xylocaine, lidoderm, 2-diethylamino-n-2,6-dimethylphenyl acetamide, anestacon, esracaine, duncaine, cappicaine, gravocain
PubChem CID 3676
ChEBI CHEBI:6456
IUPAC Name 2-(diethylamino)-N-(2,6-dimethylphenyl)acetamide
SMILES CCN(CC)CC(=O)NC1=C(C=CC=C1C)C

Specifications

Quantity 25 g
Grade Not Available
Chemical Name or Material Lidocaine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.