Learn More
Sigma Aldrich Fine Chemicals Biosciences Lidocaine powder100G
Catalog No. 502899947
Encompass
Lidocaine is a local anesthetic and an effective antiarrhythmianbspagent. It occurs as SOLUTIONons of crystalline colorless solid for injections that has a short half-life within plasma.
Catalog No. 50-289-9947 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L7757100G
Lidocaine is a local anesthetic and an effective antiarrhythmianbspagent. It occurs as SOLUTIONons of crystalline colorless solid for injections that has a short half-life within plasma.