Learn More
U.S. Pharmacopeia Lidocaine, 137-58-6, MFCD00026733, 250mg
Catalog No. NC9115960
Encompass
Molecular formula C14H22N2O, Molecular weight 234.34, Melting Point 150.8 - 158 °F (66 - 70 °C), Boiling Point 294.8 - 320 °F (146 - 160 °C), Synonyms: Lignocaine
Catalog No. NC9115960 Supplier U.S. Pharmacopeia Supplier No. 1366002
Molecular formula C14H22N2O, Molecular weight 234.34, Melting Point 150.8 - 158 °F (66 - 70 °C), Boiling Point 294.8 - 320 °F (146 - 160 °C), Synonyms: Lignocaine