Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lidocaine, USP, 97.5-102.5%, Spectrum™ Chemical
SDP

Catalog No. 18603878 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-878 5 kg Poly Pail
18-603-874 25 g Poly Bottle
18-603-875 100 g Poly Bottle
18-603-876 500 g Poly Bottle
18-603-877 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-603-878 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. LI1025KG
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LI102, 137-58-6, C14H22N2O

Spectrum™ Chemical Lidocaine, USP , also known as xylocaine or lignocaine is used as a topical local anesthetic and medication to numb tissue in a specific area. It also has uses as a treatment for ventricular tachycardia and to perform nerve blocks. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 137-58-6
Molecular Formula C14H22N2O
Molecular Weight (g/mol) 234.34
InChI Key NNJVILVZKWQKPM-UHFFFAOYSA-N
IUPAC Name 2-(diethylamino)-N-(2,6-dimethylphenyl)acetamide
SMILES CCN(CC)CC(=O)NC1=C(C)C=CC=C1C

Specifications

CAS Max % 100%
Quantity 5 kg
Packaging Poly Pail
Boiling Point 180°C to 182°C
Percent Purity 97.5 to 102.5%
Grade USP
Chemical Name or Material Lidocaine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.