Learn More
TARGETMOL CHEMICALS INC LIDORESTAT 50MG
SDP
Catalog No. 502581206
Encompass_Preferred
Also available in 5 mg 10 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lidorestat (IDD-676) is an aldose reductase inhibitor (IC50 5 nM) with effective selective and oral activity. Lidorestat improves nerve conduction and reduces the formation of cataracts. Lidorestat can be used to treat complications of chronic diabetes. purity: 99%
Catalog No. 50-258-1206 Supplier TARGETMOL CHEMICALS INC Supplier No. T1575650MG
Also available in 5 mg 10 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lidorestat (IDD-676) is an aldose reductase inhibitor (IC50 5 nM) with effective selective and oral activity. Lidorestat improves nerve conduction and reduces the formation of cataracts. Lidorestat can be used to treat complications of chronic diabetes. purity: 99%