Learn More
World Precision Instrument Liebermann Speculum, 15 mm Blades
SDP
Catalog No. 502538347
Encompass
The Leibermann Self-Retaining Wire Speculum is used in ophthalmology when examining the globe of the eye. These specula can hold the eye open during exams or procedures, giving a clear view of the eye structure. This speculum is 7 cm (2.8") long. Choose between 12 and 15 mm wide wire blades. They can open with a maximum spread of 6 cm. Made of stainless steel, these specula are sterilizable.
Catalog No. 50-253-8347 Supplier World Precision Instrument Supplier No. WP105673
The Leibermann Self-Retaining Wire Speculum is used in ophthalmology when examining the globe of the eye. These specula can hold the eye open during exams or procedures, giving a clear view of the eye structure. This speculum is 7 cm (2.8") long. Choose between 12 and 15 mm wide wire blades. They can open with a maximum spread of 6 cm. Made of stainless steel, these specula are sterilizable.