Learn More
LIEBHERR USA Upright single door, Flammable Storage scientific 4C refrigerator, 19 cuft (558 L) capacity, solid door, 115V 60 Hz
Catalog No. NC3990606
Encompass_Preferred
Upright single door, Flammable Storage scientific 4C refrigerator, 19 cuft (558 L) capacity, solid door, 115V 60 Hz, 4 adjustable glass shelves, 4 locking casters standard, field reversible door, Energy Star 2.0 Certified, DIN 13277 compliant, compliance with NFPA 45 and NFPA 99, Meets Class 1, Division 2, Gas Group B. Compliance with ATEX Directive
Catalog No. NC3990606 Supplier LIEBHERR USA Supplier No. SRFFG 5501
Upright single door, Flammable Storage scientific 4C refrigerator, 19 cuft (558 L) capacity, solid door, 115V 60 Hz, 4 adjustable glass shelves, 4 locking casters standard, field reversible door, Energy Star 2.0 Certified, DIN 13277 compliant, compliance with NFPA 45 and NFPA 99, Meets Class 1, Division 2, Gas Group B. Compliance with ATEX Directive