Learn More
TARGETMOL CHEMICALS INC LIENSININE 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T5S109925MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. 1. Liensinine a kind of isoquinoline alkaloid can antagonize the ventricular arrhythmias. purity: 99%
Catalog No. 50-257-9714