Learn More
Medchemexpress LLC HY-N0485 5mg Medchemexpress, Liensinine (Diperchlorate) CAS:5088-90-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN04855MG
Encompass_Preferred
Medchemexpress, HY-N0485 5mg Liensinine (Diperchlorate) CAS:5088-90-4 Liensinine Diperchlor​ate is a major isoquinoline alkaloid, extracted from the seed embryo of Nelumbo nucifera Gaertn. Liensinine Diperchlor​ate inhibits late-stage autophagy/mitophagy through blocking autophagosome-lysosome fusion. Liensinine Diperchlor​ate has a wide range of biological activities, including anti-arrhythmias, anti-hypertension, anti-pulmonary fibrosis, relaxation on vascular smooth muscle, etc. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0278