Learn More
Genscript Corporation LIF Bovine
Supplier:  Genscript Corporation Z038021
Encompass
LIF has the capacity to induce terminal differentiation in leukemic cells Its activities include the induction of hematopoietic differentiation in normal and myeloid leukemia cells the induction of neuronal cell differentiation and the stimulation of acute-phase protein synthesis in hepatocytes (By similarity)
Catalog No. 50-000-57524