Learn More
Medchemexpress LLC HY-P7084 10ug Medchemexpress, LIF, Mouse CAS: Purity:>98%
SDP
Catalog No. 501934323
Encompass_Preferred
Medchemexpress, HY-P7084 10ug LIF, Mouse CAS: Recombinant Mouse Leukemia Inhibitory Factor is a lymphoid factor involved in various neural and inflammatory processes such as the acute-phase reaction, tissue damage, and infection. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4323 Supplier Medchemexpress LLC Supplier No. HYP708410UG
Medchemexpress, HY-P7084 10ug LIF, Mouse CAS: Recombinant Mouse Leukemia Inhibitory Factor is a lymphoid factor involved in various neural and inflammatory processes such as the acute-phase reaction, tissue damage, and infection. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.