Learn More
Enzo Life Sciences LIF (mouse), (recombinant) (1ml)
SDP
Supplier:  Enzo Life Sciences ALX201242L001
Encompass_Preferred
Alternative name: Leukemia inhibitory factor. Purity: ≥95% (SDS-PAGE). MW: ~20kDa. Formulation: Liquid. 0.2μm-filtered solution in PBS containing 0.5% BSA. Contains no preservatives. Further dilutions should be made with medium containing 1% BSA. Source: Produced in E. coli. Long Term Storage: -20°C. Use/Stability: Stable for 6 months when stored at +4°C. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7956