Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ LIF Recombinant Mouse Protein, embryonic stem cell-qualified

Catalog No. A35935
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
100 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A35935 100 μg
A35934 50 μg
A35933 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A35935 Supplier Gibco™ Supplier No. A35935
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Gibco embryonic stem cell (ESC)–qualified mouse LIF is a recombinant protein that supports the culture and maintenance of mouse pluripotent stem cells (PSCs) by suppressing spontaneous differentiation.

Produced in barley grain to minimize endotoxin levels, each lot is carefully tested for purity, microbial contamination, bioactivity, and ability to support mouse PSC culture, thereby minimizing the risk of contamination and helping ensure robust performance.

With LIF Recombinant Mouse Protein, ESC-Qualified, you can:
• Rely on a recombinant growth factor that is qualified for mouse PSC culture
• Safeguard mouse PSC quality with a low-endotoxin LIF
• Culture with confidence with a rigorously tested reagent

Rely on an ESC-qualified growth factor
LIF bioactivity is typically measured by observing its ability to induce differentiation of murine M1 myeloid leukemia cells. In addition to this standard assay, each lot of ESC-qualified mouse LIF is tested for its ability to maintain Oct4 expression in mouse PSCs, thus helping to ensure robust performance for mouse PSC culture.

Safeguard mouse PSC quality with a low-endotoxin LIF
Unlike bacteria-derived recombinant proteins which may carry over harmful bacterial endotoxins, ESC-qualified mouse LIF is derived from plant tissue. It is verified to contain less than 0.005 ng/μg of endotoxin, more than ten times lower than most bacteria-derived counterparts. This unique, low-endotoxin LIF supports the optimal conditions for safeguarding the health and quality of your mouse PSC culture.

Culture with confidence with a rigorously tested reagent
ESC-qualified mouse LIF is tested for bacteria and mycoplasma to minimize the risk of contamination. It is analyzed by SDS-PAGE to ensure >95% purity. It is also tested for pyrogenic and pro-inflammatory contaminants via monocyte activation test.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Culture
Quantity 100 μg
Gene Symbol LIF
Species Mouse
Recombinant Recombinant
Protein Tag His Tag
Content And Storage Store in freezer (-5°C to -30°C).
Classification ES Cell-Qualified
Shipping Condition Room Temperature
Expression System Hordeum vulgare (Barley Grain)
Protein Family Cytokines & Receptors
Protein Form Recombinant
Form Lyophilized
Research Category Stem Cell Research
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
What are the differences between Leukemia Inhibitory Factor (LIF) Recombinant Mouse Protein, Embyonic Stem Cell-Qualified and LIF Recombinant Human Protein?

Besides the species difference between LIF Recombinant Mouse Protein, Embyonic Stem Cell-Qualified (Cat. Nos. A35933, A35934, A35935) and LIF Recombinant Human Protein (Cat. No. PHC9484), the other main difference is that ESC-qualified mouse LIF is derived from plant tissue, whereas LIF Recombinant Human Protein is produced in E. coli. ESC-qualified mouse LIF is verified to contain less than 0.005 ng/µg of endotoxin, which is more than ten times lower than that in E. coli-derived recombinant human LIF protein. These two recombinant proteins also have different reconstitution instructions, different storage conditions, and shelf life.

Can Leukemia Inhibitory Factor (LIF) Recombinant Mouse Protein, Embyonic Stem Cell-Qualified work with human cells?

No, Leukemia Inhibitory Factor (LIF) Recombinant Mouse Protein, Embryonic Stem Cell-Qualified cannot work in human cells as it does not bind human LIF receptor.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.