Learn More
MSC Life Latch Plastic Bucket, HDPE, Child-Resistant, Tamper-Evident
Catalog No. 502837323
Encompass
Life Latch plastic buckets are industry-standard containers for child-resistant and tamper-evident packaging. They feature a unique screw-top lid system for a moisture-resistant and airtight seal, requiring no tools. Constructed from high-performance HDPE, these UN-rated buckets are suitable for transporting hazardous chemicals, pharmaceuticals, and pool supplies, offering senior-friendliness and clear visual tampering indicators.
  • Features an industry-standard design for secure packaging
  • Incorporates a unique screw-top lid system that does not require tools
  • Provides a moisture-resistant and airtight seal
  • UN-rated for shipping various materials
  • Suitable for hazardous chemicals, pharmaceuticals, and pool supplies
  • Made from high-performance HDPE
  • Designed for child-resistance and senior-friendliness
  • Offers visual evidence of tampering
Catalog No. 50-283-7323 Supplier MSC Supplier No. 58156118
Life Latch plastic buckets are industry-standard containers for child-resistant and tamper-evident packaging. They feature a unique screw-top lid system for a moisture-resistant and airtight seal, requiring no tools. Constructed from high-performance HDPE, these UN-rated buckets are suitable for transporting hazardous chemicals, pharmaceuticals, and pool supplies, offering senior-friendliness and clear visual tampering indicators.
  • Features an industry-standard design for secure packaging
  • Incorporates a unique screw-top lid system that does not require tools
  • Provides a moisture-resistant and airtight seal
  • UN-rated for shipping various materials
  • Suitable for hazardous chemicals, pharmaceuticals, and pool supplies
  • Made from high-performance HDPE
  • Designed for child-resistance and senior-friendliness
  • Offers visual evidence of tampering