Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Basco™ Life Latch™ New Generation Bucket with Screw Top Lid, 0.6 gal.
SDP

Catalog No. 19339068
Change view
Click to view available options
Capacity (English):
0.6 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-339-068 0.6 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-339-068 Supplier Basco™ Supplier No. SS006WH
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Keep your products secured and contained in this FDA-compliant bucket with screw on lid.

Keep products secure with this 0.6 gallon bucket with screw on lid and a unique trigger mechanism to automatically secure the lid when closing.

  • Built-in plastic handle for convenient carrying
  • Removable and reusable plastic cover
  • FDA compliant food storage container
  • Easy finger activated release for easy opening of lid
  • Senior friendly and child resistant
  • UN Rated screw top plastic containers for hazardous materials

Specifications

Closure Included Yes
Closure Type Lid
Capacity (English) 0.6 gal.
Capacity (Metric) 2.3 L
Color White
Material HDPE
Shape Round
Width (English) 0.166 lb.
Height (English) 7.39 in.
Diameter (English) Opening 6.06 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.