Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Basco™ Life Latch™ New Generation Pail with Screw Top Lid, 2.5 gal.

Description
Keep products secure with this 2.5 gallon bucket with screw on lid and a unique trigger mechanism to automatically secure the lid when closing.
- Built-in plastic handle for convenient carrying
- Removable and reusable plastic cover
- FDA compliant food storage container
- Easy finger activated release for easy opening of lid
- Senior friendly and child resistant
- UN Rated screw top plastic containers for hazardous materials
Specifications
Specifications
| Closure Included | Yes |
| Closure Type | Lid |
| Capacity (English) | 2.5 gal. |
| Capacity (Metric) | 9.5 L |
| Color | White |
| Material | HDPE |
| Shape | Round |
| Height (English) | 9.8 in. |
| Diameter (English) Opening | 10.08 in. |
| Weight (English) | 0.76 lb. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?