Learn More
StatLab Life Latch New Generation Specimen Containers
SDP
Catalog No. 21010344
Change view
Click to view available options
Quantity:
2.5 gal.
3.4 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-010-344 2.5 gal.
22-021-453 3.4 gal.
22-021-455 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 21-010-344 Supplier StatLab Supplier No. MM2.5GAL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed for safe, leak-resistant transport and storage of fluids.
  • Screw-top containers have knurled cap and base for easy handling
  • Containers are graduated and made of durable clarified polypropylene
Specifications
Product Type Specimen Container
Closure Included Yes
Graduated Yes
Material Polypropylene
Shape Cylindrical
Sterility Non-sterile
No. per Case 8/Cs.
Quantity 2.5 gal.