Learn More
StatLab Life Latch New Generation Specimen Containers
Description
- Screw-top containers have knurled cap and base for easy handling
- Containers are graduated and made of durable clarified polypropylene
Specifications
Specifications
| Product Type | Specimen Container |
| Closure Included | Yes |
| Graduated | Yes |
| Material | Polypropylene |
| Shape | Cylindrical |
| Sterility | Non-sterile |