Learn More
LIFE™ OxygenPac™ Emergency Oxygen in Wall Case
Catalog No. 17908 Shop All Life Corporation Products
Click to view available options
Product Type:
SoftPac Emergency Oxygen Unit
Catalog No. Product Type
17-908 SoftPac Emergency Oxygen Unit
1 options
Catalog No. 17-908 Supplier LIFE™ Supplier No. LIFE6FF
For industrial and office workplace first aid programs, LIFE™ OxygenPac™ is simple and easy to use to give crucial lifesaving oxygen until medical help arrives.
  • Administer oxygen easily and quickly with unit featuring transparent cover and clear two-step instructions.
  • Serves as inhalator for breathing victim, resuscitator for nonbreathing victim. For resuscitation, mask with one-way valve eliminates direct contact, prevents transfer of body fluids.
  • Convenient handle for wall mounting or transport.
  • Refillable tank holds 90-minute supply of oxygen at fixed flowrate of 6L/min. for 45% inspiration 02 concentration to provide recommended 100% inspired O2.
  • Unit measures 9L x 6 3/8W x 20 in.H (23 x 16 x 51cm); weighs 16 lb. (7kg).
  • Unit includes O2 fill and fasteners for wall mounting.
  • Durable case, clear cover
  • Two-step instructions, simple on/off lever, constant reading supply gague
  • Universally fits both adults and children
Specifications
Product Type SoftPac Emergency Oxygen Unit
Material Aluminum
For Use With (Application) Serves as inhalator for breathing victim, resuscitator for nonbreathing victim
Includes O2 fill and fasteners for wall mounting
Quantity 1 Ea.
Weight (English) 16 lb.
Weight (Metric) 7 kg