Learn More
Advanced Biomatrix LifeSupport Support Slurry for FRESH Bioprinting
Catalog No. NC2033810
Encompass
LifeSupport is the industry standard for FRESH bioprinting. FRESH stands for Freeform Reversible Embedding of Suspended Hydrogels. Each LifeSupport printing kit comes with 5 units of 2 grams of sterile LifeSupport powder. Simply resuspend, centrifuge and print. LifeSupport is the core component and industry standard for FRESH bioprinting. FRESH stands for Freeform Reversible Embedding of Suspended Hydrogels. Each LifeSupport printing kit comes with 5 units of 2 grams of sterile LifeSupport powder. One gram of powder hydrates to approximately 10 mL of FRESH support material.
Catalog No. NC2033810 Supplier Advanced Biomatrix Supplier No. 524410GM
LifeSupport is the industry standard for FRESH bioprinting. FRESH stands for Freeform Reversible Embedding of Suspended Hydrogels. Each LifeSupport printing kit comes with 5 units of 2 grams of sterile LifeSupport powder. Simply resuspend, centrifuge and print. LifeSupport is the core component and industry standard for FRESH bioprinting. FRESH stands for Freeform Reversible Embedding of Suspended Hydrogels. Each LifeSupport printing kit comes with 5 units of 2 grams of sterile LifeSupport powder. One gram of powder hydrates to approximately 10 mL of FRESH support material.