Learn More
SOLVENTUM 3M™ LifeASSURE™ PDA Series Filter Capsule
Supplier:  SOLVENTUM 7000051535
Encompass_Preferred
Validated 0.2 and 0.1 micron rated membrane provides reliable sterilizing filtration. Excellent contaminant capacity, flow rates, and life resulting from robust double-asymmetric membrane layer filter construction. Hydrophilic asymmetric polyethersulfone membrane enables wet out in aqueous solutions for integrity testing. Advanced Pleat Technology construction increases the useful surface area by increasing the flow paths between media pleats.
Catalog No. NC1550772