Learn More
Mfi Medical LIFEDOP 250 ABI SYSTEM
SDP
Catalog No. NC3719571
Encompass_Preferred
  • Portable bi-directional Doppler system for accurate diagnosis of Peripheral Arterial Disease (P.A.D.)
  • Handheld system enables easy vessel location and blood flow detection
  • Measures systolic pressures for ABI calculation
  • Includes compact rechargeable printer for waveform capture
  • Printout on adhesive-backed label paper for compliance documentation
  • Automatic scaling and calibration for reliable performance
  • LCD display shows battery status, blood flow direction, and relative frequency
  • Power source options: 3 x AA batteries or lithium ion
  • Forty 12-minute exams per battery charge
  • Dimensions: 147 mm H x 70 mm W x 35 mm L
  • Weight: 320 g
  • 1 year manufacturer warranty
  • CPT Reimbursement Code: 93922
  • Includes carrying case and examination guides
Catalog No. NC3719571 Supplier Mfi Medical Supplier No. WLCL250AB
  • Portable bi-directional Doppler system for accurate diagnosis of Peripheral Arterial Disease (P.A.D.)
  • Handheld system enables easy vessel location and blood flow detection
  • Measures systolic pressures for ABI calculation
  • Includes compact rechargeable printer for waveform capture
  • Printout on adhesive-backed label paper for compliance documentation
  • Automatic scaling and calibration for reliable performance
  • LCD display shows battery status, blood flow direction, and relative frequency
  • Power source options: 3 x AA batteries or lithium ion
  • Forty 12-minute exams per battery charge
  • Dimensions: 147 mm H x 70 mm W x 35 mm L
  • Weight: 320 g
  • 1 year manufacturer warranty
  • CPT Reimbursement Code: 93922
  • Includes carrying case and examination guides