Learn More
Advanced Biomatrix Lifeink 220 Type I Collagen Bioink, 70 mg/ml, pH neutral
Supplier:  Advanced Biomatrix 53434ML
Encompass_Preferred
Lifeink 220 is a 70 mg/ml type I collagen bioink for extrusion based FRESH 3D bioprinting. Lifeink 220 comes as a pH neutral, isotonic bioink and is ready for cell addition and bioprinting.
Catalog No. 50-225-5940