Learn More
Advanced Biomatrix Lifeink 260 Type I Collagen Bioink, 70 mg/ml, acidic 4 mL
Catalog No. 502255954
Encompass_Preferred
Lifeink 260 is a 70 mg/ml type I collagen bioink for extrusion based FRESH 3D bioprinting. Lifeink 260 comes with acidic pH and when FRESH printed into LifeSupport yields a neutralized, high resolution collagen scaffold.
Catalog No. 50-225-5954 Supplier Advanced Biomatrix Supplier No. 53584ML
Lifeink 260 is a 70 mg/ml type I collagen bioink for extrusion based FRESH 3D bioprinting. Lifeink 260 comes with acidic pH and when FRESH printed into LifeSupport yields a neutralized, high resolution collagen scaffold.