Learn More
StatLab LifeLatch Empty Specimen Containers
SDP
Catalog No. 21033447
Change view
Click to view available options
Color:
White
Catalog No. Color
21-033-447 White
1 options
Catalog No. 21-033-447 Supplier StatLab Supplier No. MM1.25GAL
Add to Cart
Edge
Add to Cart
Designed for safe, leak-resistant transport and storage of fluids
  • Screw-top containers have knurled cap and base for easy handling
  • Graduated and made of HDPE
Specifications
Capacity (English) 1.25 gal.
Color White
Autoclavable Not Autoclavable
Material HDPE
Product Type Specimen Container