Learn More
LifeScan™ SureStep™ System
Catalog No. 23029306
Change view
Click to view available options
Surestep Control Solutn 2vl/pk
Surestep Pro Controls 3/pk
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-029306 Surestep Control Solutn 2vl/pk
23-029305 Surestep Pro Controls 3/pk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-029306 Supplier LifeScan™ Supplier No. 53885063702
This item is not returnable. View return policy
Includes
Kit Contents:
  • SureStep™
  • Meter
  • 25 SureStep™
  • Test Strips
  • PENLET™
  • II Automatic Blood Sampler
  • 25 Lancets
  • Logbook
  • Two AAA Batteries
  • Carrying Case
  • Control Solution
  • Owners Manual†
Designed for personal or bedside use
  • Stores up to 150 test results
  • Results in 15 seconds
  • Glucose range: 0 to 500mg/dL; hematocrit range: Adult 25 to 60%
  • Offers testing date/time stamp and QC event flag
  • Extra-large display
  • Ten-test memory
  • Blue confirmation dot indicates adequate sample application
† Includes SureStep™ Meter, operating guide and quick reference manual
Specifications
Product Type Surestep Control Solution
For Use With (Equipment) Sure step system
Quantity 2