Learn More
Sigma Aldrich Fine Chemicals Biosciences LifeSep magnetic separatio
Catalog No. 502891570
Encompass
Magnetic separators for single tubes are durable and easy-to-use. For use in cell sorting, RNA and DNA isolation and purification of biomolecules. Fast and easy aspiration is achieved with our Neodymium-Iron-Boron (Nd-Fe-B) magnet configuration. Patented Nd-Fe-B magnet configuration for optimal resultsOpen-faced design for clear visibilityRapid separation of micro- and nanometer beadsEasy aspirationSuperior particle retentionRemovable base for easy cleaningDurable and easy to use
Catalog No. 50-289-1570 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7401554EA
Add to Cart
Add to Cart
Magnetic separators for single tubes are durable and easy-to-use. For use in cell sorting, RNA and DNA isolation and purification of biomolecules. Fast and easy aspiration is achieved with our Neodymium-Iron-Boron (Nd-Fe-B) magnet configuration. Patented Nd-Fe-B magnet configuration for optimal resultsOpen-faced design for clear visibilityRapid separation of micro- and nanometer beadsEasy aspirationSuperior particle retentionRemovable base for easy cleaningDurable and easy to use